Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
50 mL | Estimated to ship on06 November 2026fromAreál Kühne+Nagel spol. s r.o. | 2 430,00 CZK |
About This Item
technique(s)
immunohistochemistry: suitable, western blot: suitable
General description
Application
To 9 mL of deionized water add 1 mL of ES010 and 0.1 mL H2O2. This can then be used to dilute ES005. No preservatives added.
Physical form
Preparation Note
Disclaimer
1 of 1
This Item | |
|---|---|
| description - | description Protein-Concentrate Kit (Micro) is suitable for concentrating proteins for running gels, raising antibodies, protein purification, protein assays & other applications. |
| technique(s) immunohistochemistry: suitable, western blot: suitable | technique(s) protein purification: suitable (concentration) |
Storage Class
12 - Non Combustible Liquids
WGK
WGK 2
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
77-92-9
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

