Skip to Content
Merck

FR139398085

K-020 Red Std PS 10%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene

Quality Segment

form

microspheres

concentration

10.0%

description

Functional Group Plain, <5% (Standard Deviation)

diam.

0.176-0.225 μm

color

red

Packaging

Polyethylene bottle


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library