Skip to Content
Merck

Skip To

MABC155

Anti-KIF3A Antibody, clone 4F9.1

clone 4F9.1, from mouse

Synonym(s):

Microtubule plus end-directed kinesin motor 3A, KIF3A, Kinesin-like protein KIF3A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship on03 December 2026fromAreál Kühne+Nagel spol. s r.o.

11 200,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
4F9.1, monoclonal
Application:
IHC, WB
Citations:
-

11 200,00 CZK


Estimated to ship on03 December 2026Details


​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4F9.1, monoclonal

species reactivity

rat, mouse, human

technique(s)

immunohistochemistry: suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... KIF3A(11127)

General description

Kinesin-like protein KIF3A (UniProt Q9Y496; also known as Kinesin family protein 3A, Microtubule plus end-directed kinesin motor 3A) is encoded by the KIF3A (also known as FLA10, KIF3, KLP-20) gene (Gene ID 11127) in human. Kinesin-2 is a heterotrimeric motor protein composed of two kinesin-related subunits, KIF3A and KIF3B, and an associated protein KAP3. The kinesin-2 motor KIF3A/B drives intraflagellar transport (IFT) of tubulins and other axonemal building blocks in cilia and flagella, pigment granules in melanosomes, and vesicles in neurons. Defects in these transport processes are linked to polycystic kidney disease and neurodegenerative diseases.
~75 kDa observed

Immunogen

Epitope: C-terminal
KLH-conjugated linear peptide corresponding to the C-terminal of human KIF3A.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-KIF3A Antibody, clone 4F9.1 is validated for use in Western Blotting and Immunohistochemistry for the detection of KIF3A.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected KIF3A in 10 µg of rat hippocampus tissue lysate.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected KIF3A in human cerebellum and human cerebral cortex tissue.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in mouse cerebellum tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected KIF3A in 10 µg of mouse cerebellum tissue lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
K3513SAB5702608AV33868
description

clone 4F9.1, from mouse

description

IgG fraction of antiserum, buffered aqueous solution

description

-

description

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

clone

4F9.1, monoclonal

clone

polyclonal

clone

1C8K4, monoclonal

clone

polyclonal

Gene Information

human ... KIF3A(11127)

Gene Information

human ... KIF3A(11127)
mouse ... Kif3a(16568)
rat ... Kif3a(84392)

Gene Information

human ... KIF3A(11127)

Gene Information

human ... KIF5A(3798)

species reactivity

rat, mouse, human

species reactivity

rat, mouse, human

species reactivity

rat, human

species reactivity

rabbit, dog, human

antibody form

purified antibody

antibody form

IgG fraction of antiserum

antibody form

-

antibody form

affinity isolated antibody

biological source

mouse

biological source

rabbit

biological source

rabbit

biological source

rabbit


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters