Skip to Content
Merck

Skip To

MABC563

Anti-Kallikrein 2/KLK2 Antibody, clone 4F2.3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship on26 October 2026fromAreál Kühne+Nagel spol. s r.o.

11 900,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
4F2.3, monoclonal
Application:
IHC, WB
Citations:
-

11 900,00 CZK


Estimated to ship on26 October 2026Details


​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

unconjugated

Quality Segment

antibody form

purified antibody

clone

4F2.3, monoclonal

purified by

using protein G

species reactivity

mouse, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunohistochemistry: suitable, western blot: suitable

isotype

IgG1κ

UniProt accession no.

target post-translational modification

unmodified

General description

GST-tagged recombinant protein corresponding to human Kallikrein 2/KLK2.
~30 kDa observed. Uncharacterized band(s) may appear in some lysates.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Kallikrein 2/KLK2 in human prostate tissue.

Physical form

Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in LNCaP cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Kallikrein 2/KLK2 in 10 µg of LNCaP cell lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABC971ABC470MABC552
description

-

description

clone 33.1, from mouse

description

from rabbit, purified by affinity chromatography

description

clone 11C7.1, from mouse

clone

4F2.3, monoclonal

clone

33.1, monoclonal

clone

polyclonal

clone

11C7.1, monoclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

mouse, human

species reactivity

human

species reactivity

mouse, human

species reactivity

human

antibody form

purified antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

purified antibody

technique(s)

immunohistochemistry: suitable, western blot: suitable

technique(s)

ELISA: suitable, immunohistochemistry: suitable

technique(s)

immunohistochemistry: suitable, western blot: suitable

technique(s)

western blot: suitable, immunohistochemistry: suitable (paraffin)

UniProt accession no.

P20151

UniProt accession no.

Q9UKR3

UniProt accession no.

Q9Y337

UniProt accession no.

P06870




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library