Skip to Content
Merck

Skip To

MABC960

Anti-CD1d Antibody, clone WTH-2

clone WTH-2, from mouse

Synonym(s):

Antigen-presenting glycoprotein CD1d, CD1d

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Please contact Customer Service for Availability
13 100,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
WTH-2, monoclonal
Application:
FACS, IP, WB, activity assay
Citations:
-

13 100,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

WTH-2, monoclonal

species reactivity

rat, mouse

technique(s)

activity assay: suitable, flow cytometry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Cd1D(110290701)
rat ... Cd1D(104874051)

General description

CD1D antigen, CD1d1 antigen, T-cell surface glycoprotein CD1d) is encoded by the Cd1d (also known as Cd1d1) gene (Gene ID 25109) in rat species. CD1d is a nonpolymorphic MHC class I-like molecule which presents nonpeptide ligands (e.g. glycolipids) to natrual killer T (NKT) cells. NKT cells are known to have multiple effects on innate and adaptive immune responses and on the development of pathological conditions. In additon to hematopoietic cells, CD1d protein is also reported to be detected in non-lymphatic tissues in rat, such as the exocrine pancreas and Paneth cells. CD1d is initially produced with a signal peptide sequence (a.a. 1-17), the removal of which yields the mature CD1d with a large extracellular region (a.a.18-304), a transmembrane domain (a.a. 305-325), and a short cytoplasmic tail (a.a. 326-336).
~38 kDa calculated

Immunogen

Epitope: extracellular domain
Rat CD1d-transduced M12.4.1.C3 cells.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-CD1d Antibody, clone WTH-2 is validated for use in Flow Cytometry, Western Blotting, Immunoprecipitation, and Activity Assay for the detection of CD1d.
Western Blotting Analysis: A representative lot detected exogenously expressed murine and rat CD1d in lysates of transfected Raji cells (Monzon-Casanova, E., et al. (2010). PLoS One.;5(9): e13089).
Flow Cytometry Analysis: A representative lot detected the surface expression of CD1d on isolated thymocytes from rats and mice, as well as on Raji transfectants expressing murine or rat CD1d (Monzon-Casanova, E., et al. (2010). PLoS One. 5(9): e13089).
Immunoprecipitation Analysis: A representative lot immunoprecipitated CD1d from isolated mouse and rat thymocytes, as well as exogenously expressed murine and rat CD1d from transfected Raji cells (Monzon-Casanova, E., et al. (2010). PLoS One. 5(9): e13089).
Activity Assay: A representative lot inhibited CD1d antigen presentation cells from activating type I NK cells using total splenocytes from rats and mice. Clone WTH-2 also blocked mCD1d1-transduced LBB cells from activating co-cultured VIII24.1 murine type II NKT cells, as well as mCD1d1-transduced Raji cells from activating co-cultured XV19.2 T murine type II NKT cells (Monzon-Casanova, E., et al. (2010). PLoS One. 5(9): e13089).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in PBS without preservatives.

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Flow Cytometry in rat thymocytes and splenocytes.

Flow Cytometry Analysis: 1.0 µg of this antibody detected CD1d in rat thymocytes and splenocytes.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABC959MABF3243ZMS1223
description

clone WTH-2, from mouse

description

clone WTH-1, from mouse

description

-

description

recombinant, expressed in HEK 293 cells

clone

WTH-2, monoclonal

clone

WTH-1, monoclonal

clone

CD1d42, monoclonal

clone

51.1, recombinant monoclonal

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

species reactivity

rat, mouse

species reactivity

mouse, rat

species reactivity

human

species reactivity

human

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Gene Information

mouse ... Cd1D(110290701)
rat ... Cd1D(104874051)

Gene Information

mouse ... Cd1D(110290701)
rat ... Cd1D(104874051)

Gene Information

human ... CD1D(912)

Gene Information

human ... CD1D(912)

antibody form

purified immunoglobulin

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters