Skip to Content
Merck

Skip To

MCGEXPRIN

MAS-100 CG EX® External Printer

for use with MAS-100 CG Ex® (microbial compressed gas sampler)

Synonym(s):

External Printer, Printer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Check Cart for Availability
52 300,00 CZK

About This Item

NACRES:
JA.11
UNSPSC Code:
41103403

52 300,00 CZK


Check Cart for Availability

​
Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

MAS-100 CG Ex®

application(s)

cosmetics
food and beverages
pharmaceutical

compatibility

for use with MAS-100 CG Ex® (microbial compressed gas sampler)

General description

MAS-100 CG Ex® External Printer is designed to print the sampling parameters of MAS-100 CG Ex® microbial compressed gas sampler after sampling of compressed gases like compressed air, oxygen, nitrogen etc.

Other Notes

Please note the MAS-100 CG Ex® External Printer comes with compatible cable.

Legal Information

MAS-100 CG Ex is a registered trademark of MBV AG




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters