Skip to Content

System maintenance is scheduled for Sunday, October 11, 8:00 AM to 12:00 PM CST (Sunday, 11th October 2026, 03:00 PM to 07:00 PM CET). Quotes, price checks, and order status inquiries may be temporarily unavailable.

Merck

Key Documents

Skip To

MX0095

Malachite Green Oxalate

90% (TiCl3, titration), Amax, 616-620 nm 0.51

Synonym(s):

Malachite Green Oxalate, Basic Green 4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
2[NH(CH3)2C6H4C(C6H5)=C6H4=N(CH3)2]+3HOOCCOOH
CAS Number:
Colour Index Number:
42000
UNSPSC Code:
41116121
EC Index Number:
241-922-5
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

90% (TiCl3, titration)

form

crystals

color

green

mp

~159 °C

Amax

0.51 at 616-620 nm

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

15-25°C

SMILES string

N(C)(C)c1ccc(cc1)[C+](c3ccc(cc3)N(C)C)c2ccccc2

InChI

1S/C23H25N2/c1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4/h5-17H,1-4H3/q+1

InChI key

VFCNQNZNPKRXIT-UHFFFAOYSA-N

General description

For staining of bacterial polysaccharides and spores
Malachite Green Oxalate
MX0095-3 25g Glass Bottle

Application

For In-Vitro Diagnostic Use.
For Professional Use.
For staining of bacterial polysaccharides and spores

Analysis Note

Absorbance at maximum0.51AU minAbsorbance maximum wavelength616-620nmAbsorbance ratio; (P-15/P+15)1.00-1.15Assay (TiCl3 titration)90% minColorGreenFormCrystals


Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Regarding MX0095-3 on the SDS it use to carry the Molecular weight and now it does not. I have 2 questions: 1. why does the SDS not have this molecular weight listed anymore and 2. when was this material discontinued?

    1 answer
    1. This product has been deleted for over 1 year. Other Malachite Green oxalate salt products remain available. The molecular weight for this product is not published on the product page or other documents. However, it is reported under the CAS number at PubChem. See the links below for additional information:

      PubChem CAS 18015-76-4
      https://pubchem.ncbi.nlm.nih.gov/compound/14513293

      Other Malachite Green product options
      https://www.sigmaaldrich.com/search/malachite-green-oxalate-salt?focus=products&page=1&perpage=30&sort=relevance&term=malachite%20green%20oxalate%20salt&type=product

      Helpful?

Reviews

No rating value

Active Filters