Skip to Content
Merck

Skip To

MX0335

Mercuric Acetate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Mercuric Acetate, Acetic Acid,Mercuric Salt,Mercury(II) Acetate, Acetic Acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(CH3COO)2Hg
CAS Number:
EC Index Number:
216-491-1
UNSPSC Code:
12352103
MDL number:
eCl@ss:
32039090
Assay:
≥99.0% (complexometric)
Grade:
ACS reagent
Form:
solid
Solubility:
400 g/L
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

assay

≥99.0% (complexometric)

form

solid

reduction residue

≤0.02%

mp

178-180 °C (decomposition)

solubility

400 g/L

anion traces

chloride (Cl-): ≤0.005%, nitrate (NO3-): ≤0.005%, sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%, Hg+: ≤0.3%, heavy metals: ≤0.002%

shipped in

ambient

storage temp.

room temp

InChI

1S/2C2H4O2.Hg/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

BRMYZIKAHFEUFJ-UHFFFAOYSA-L

Application

Contains mercury. Do not put in trash. Manage as hazardous waste.

Analysis Note

Assay (complexometric): 99.0% min
Chloride (Cl): 0.005% max
Iron (Fe): 0.001% max
Mercury (I) (as Hg): 0.3% max
Nitrate (NO3): 0.005% max
Other heavy metals: 0.002% max
Residue after reduction: 0.02% max
Substances insoluble in dilute acetic acid: %
Sulfate (SO4): 0.005% max

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.


Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

1600-27-7

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters