The Milliflex fluidic hub is VHP compatible. It can be used for pharmaceutical product analysis, bioburden testing, water monitoring and sterility testing.
description
-
compatibility
standard for VHP, for use with MILLIFLEX®
compatibility
for use with Milliflex®
compatibility
for use with MILLIFLEX®, standard for VHP
compatibility
for use with MILLIFLEX®, standard for VHP
shipped in
ambient
shipped in
ambient
shipped in
ambient
shipped in
ambient
input
sample type pharmaceutical(s)
input
-
input
sample type pharmaceutical(s)
input
pharmaceutical(s) sample type pharmaceutical(s)
technique(s)
Rapid Sterility Testing: suitable
technique(s)
-
technique(s)
Rapid Sterility Testing: suitable
technique(s)
Rapid Sterility Testing: suitable
manufacturer/tradename
PyroDetect, Milliflex®
manufacturer/tradename
Milliflex Oasis®
manufacturer/tradename
Milliflex®
manufacturer/tradename
Milliflex®
Quality Level
100
Quality Level
SP2
Quality Level
100
Quality Level
-
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.