Skip to Content
Merck

Skip To

MXPTUBVHP

VHP tubes set for Milliflex fluidic hub

The VHP tube set for the Milliflex fluidic hub is usable for pharmaceutical product analysis, bioburden testing and water monitoring.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
23151818
NACRES:
NB.79
eCl@ss:
32014001
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

manufacturer/tradename

Milliflex®

technique(s)

Rapid Sterility Testing: suitable

input

sample type pharmaceutical(s)
pharmaceutical(s)

application(s)

bioburden testing
pharmaceutical
sterility testing
water monitoring

compatibility

for use with Milliflex®, standard for VHP

shipped in

ambient

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
VHPHEADMMMXPFHUVHPMXPELCVHP
description

The VHP tube set for the Milliflex fluidic hub is usable for pharmaceutical product analysis, bioburden testing and water monitoring.

description

for use with Milliflex®, Standard For VHP

description

The Milliflex fluidic hub is VHP compatible. It can be used for pharmaceutical product analysis, bioburden testing, water monitoring and sterility testing.

description

-

compatibility

standard for VHP, for use with Milliflex®

compatibility

for use with Milliflex®

compatibility

standard for VHP, for use with MILLIFLEX®

compatibility

for use with MILLIFLEX®, standard for VHP

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

Quality Level

100

Quality Level

SP2

Quality Level

100

Quality Level

100

input

sample type pharmaceutical(s)
pharmaceutical(s)

input

-

input

sample type pharmaceutical(s)

input

pharmaceutical(s)
sample type pharmaceutical(s)

technique(s)

Rapid Sterility Testing: suitable

technique(s)

-

technique(s)

Rapid Sterility Testing: suitable

technique(s)

Rapid Sterility Testing: suitable

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex Oasis®

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex®




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters