Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12352203
Pricing and availability is not currently available.
biological source
rabbit
Quality Segment
antibody product type
primary antibodies
form
liquid
does not contain
preservative
species reactivity
human
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, avoid repeated freeze/thaw cycles
isotype
IgG
shipped in
wet ice
storage temp.
−20°C
General description
Rabbit polyclonal antibody supplied as undiluted serum. Recognizes the ~53 kDa p53 phoshorylated at Ser15.
This PhosphoDetect Anti-p53 (pSer¹⁵) (Ab-6) Rabbit pAb is validated for use in Immunoblotting, Paraffin Sections for the detection of p53 (pSer¹⁵) (Ab-6).
This product has been discontinued.
Recognizes the ~53 kDa p53 protein phosphorylated at Ser15. Does not recognizes unphosphorylated p53.
Recognizes the ~53 kDa p53 protein phosphorylated at Ser15. Does not recognizes unphosphorylated p53.
Immunogen
Human
a synthetic phosphopeptide [VEPPLpSQETFS(C)] corresponding to amino acids 10-20 surrounding the Ser¹⁵ phosphorylation site of human p53
Application

Immunoblotting (1:5000-1:10,000)
Paraffin Sections (1:1000-1:3000, heat pre-treatment required)
Physical form
Undiluted serum.
Preparation Note
Following initial thaw, aliquot and freeze (-20°C).
Analysis Note
Positive Control
MCF7 cells or prostate carcinoma tissue
MCF7 cells or prostate carcinoma tissue
Other Notes
Does not recognizes unphosphorylated p53. Antibody should be titrated for optimal results in individual systems.
Levine, A.J. 1997. Cell88, 323.
Meek, D.W. 1997. Pathol. Biol. Paris45, 804.
Miller, S.D., et al. 1997. Mol. Cell. Biol.17, 2194.
Pellegata, N.S., et al. 1996. Proc. Natl. Acad. Sci. USA93, 15209.
Sturzbecher, H.-W., et al. 1996. EMBO J.15, 1992.
Argarwal, M.L., et al. 1995. Proc. Natl. Acad. Sci. USA92, 8493.
Götz, C. and Montenarh, M. 1995. Int. J. Oncology6, 1129.
Reed, M. et al. 1995. Proc. Natl. Acad. Sci. USA92, 9455.
Waldman, T., et al. 1995. Cancer Res.55, 5187.
Cho, Y., et al. 1994. Science265, 346.
Clarke, A.R., et al. 1994. Oncogene9, 1767.
El-Deiry, W.S., et al. 1994. Cancer Res.54, 1169.
Greenblatt, M.S., et al. 1994. Cancer Res.54, 4855.
Lane, D.P. 1992. Nature358, 15.
Meek, D.W. 1997. Pathol. Biol. Paris45, 804.
Miller, S.D., et al. 1997. Mol. Cell. Biol.17, 2194.
Pellegata, N.S., et al. 1996. Proc. Natl. Acad. Sci. USA93, 15209.
Sturzbecher, H.-W., et al. 1996. EMBO J.15, 1992.
Argarwal, M.L., et al. 1995. Proc. Natl. Acad. Sci. USA92, 8493.
Götz, C. and Montenarh, M. 1995. Int. J. Oncology6, 1129.
Reed, M. et al. 1995. Proc. Natl. Acad. Sci. USA92, 9455.
Waldman, T., et al. 1995. Cancer Res.55, 5187.
Cho, Y., et al. 1994. Science265, 346.
Clarke, A.R., et al. 1994. Oncogene9, 1767.
El-Deiry, W.S., et al. 1994. Cancer Res.54, 1169.
Greenblatt, M.S., et al. 1994. Cancer Res.54, 4855.
Lane, D.P. 1992. Nature358, 15.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10-13 - German Storage Class 10 to 13
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.