Skip to Content
Merck

SCC239

YUMM 4.3 Mouse Melanoma Cell Line

Synonym(s):

YUMM 4.3 Mouse Melanoma Cell Line, Yale University Mouse Melanoma 4.3 cell line, Yale University Mouse Melanoma cell line

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
NACRES:
NA.51
eCl@ss:
32040201
Biological source:
mouse
Manufacturer/tradename:
Millipore
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

packaging

vial of ≥1X10⁶ vial (viable cells per vial)

manufacturer/tradename

Millipore

technique(s)

cell culture | mammalian: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

General description

As immune-based therapies in cancer models become more popular and applicable, a further need for immune-competent models has been presented. A wide variety of genetically engineered mouse models have been created to express vital melanoma genotypes. The variety in these models have allowed the study of genetic alterations that are relevant to human melanoma biology. While it is quite useful to have mouse models with genetically defined human melanoma characteristics, itis limited by complex mouse colony maintenance.

The YUMM4.2 cell line was produced to be more experimentally relevant. The cell line was derived by backcrossing important alleles into C57BL/6J mice. The mice were interbred to produce human-relevant genetically defined mouse models that could then be used to generate cell lines. Melanomas were induced by applying 4-hydroxytamoxifen to induce Cre-lox allele recombination. Cells were then mechanically and enzymatically dissociated from the removed tumor. The melanoma lines generated from this procedure retained important human melanoma characteristics useful for studying disease progression.

The YUMM4.2 cell line is derived from a female mouse model driven by Pten inactivation and Cdkn2a inactivation. The cell line is positive for the common melanoma markers, MelanA and PMEL. The cells are syngeneic with C57BL/6J mice and are very tumorigenic in vivo.

References:
1. Meeth K et al. (2016) The YUMM lines: a series of congenic mouse melanoma cell lines with defined genetic alterations. Pigment Cell Melanoma Res 29(5): 590-597.

Application

  • Each vial contains > 1X106 viable cells.
  • Cells are tested negative for infectious diseases by a Mouse Essential CLEAR panel by Charles River Animal Diagnostic Services.
  • Cells are verified to be of mouse origin and negative for inter-species contamination from rat, chinese hamster, Golden Syrian hamster, human and non-human primate (NHP) as assessed by a Contamination CLEAR panel by Charles River Animal Diagnostic Services.
  • Cells are negative for mycoplasma contamination.

Features and Benefits

YUMM4.2 cell line is derived from a female mouse model driven by Pten inactivation and Cdkn2a inactivation. The cell line is positive for the common melanoma markers, MelanA and PMEL.

Preparation Note

YUMM 4.3 cells should be stored in liquid nitrogen until use. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Analysis Note

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@milliporesigma.com.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library