Skip to Content
Merck

Skip To

SCC482

Replic Mouse Renal Cell Line

Synonym(s):

Replic Mouse Renal Cell Line, REP cell-lineage immortalized and cultivable, REP cells

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81
Biological source:
mouse
Relevant disease(s):
cancer
Technique(s):
cell culture | mammalian: suitable
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

description

cell culture mammalian: suitable

packaging

vial of ≥1X10⁶ vial (viable cells per vial)

manufacturer/tradename

Millipore

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

liquid nitrogen

storage temp.

−196°C

General description

Chronic kidney disease (CKD) is caused by a variety of health factors but is often characterized by improper kidney function or abnormal kidney structure. Patients often start out asymptomatic but develop symptoms later in disease progression. Erythropoietin (Epo) is a hormone produced by the kidneys to induce red blood cell production in the body. Due to epo′s direct connection to the functionality of the kidneys, epo-deficiency anemia is a common problem that arises from CKD. Another complication that commonly affects CKD patients is renal fibrosis, a condition in which the kidneys become further damaged due to the excess production of extracellular matrix (ECM). This excess ECM production originates in myofibroblasts found in the peritubular interstitia. These dangerous myofibroblasts most likely originate through the transformation of tubular epithelial cells, capillary endothelial cells, migratory bone marrow cells, and interstitial fibroblasts. Interstitial fibroblasts are thought to be a major contributor due to the inflammation signaling response activated by nephron injury. However, as interstitial fibroblasts undergo myofibroblastic transformation, epo-production capabilities are inactivated. The general lack of appropriate cell models has made it difficult to understand epo-producing cells which also hinders the ability to understand underlying mechanisms of renal fibrosis. The SCC482 Replic Mouse Renal Cell line represents an important cell model in understanding the molecular pathophysiology of epo-producing cells. The cell line is derived from mouse renal epo-producing cells which were isolated and immortalized by exogenous expression of oncogenic H-RAS. The cells retain tdTomato expression which was utilized for isolation and cultivation. However, the Replic cells have lost their ability to produce erythropoietin due to continuous DNA methylation in the Epo gene promoter. The unique myofibroblastic features such as high TGFβ and the epo-producing defect of the cell line allows for a more thorough understanding of kidney fibrosis, renal anemia, and molecular CKD pathology.SourceDerived from kidneys of ISAM-REC mice which were later infected with lentivirus vector expressing G12V oncogenic mutant of human H-RAS. Cells were positively selected using tdTomato fluorescent expression.References1. Sato K, Hirano I, Sekine H, Miyauchi K, Nakai T, Kato K, Ito S, Yamamoto M, Suzuki N. 2019. An immortalized cell line derived from renal erythropoietin-producing (REP) cells demonstrates their potential to transform into myofibroblasts. Scientific Reports. 9(1).

Application

  • Each vial contains > 1X106 viable cells.
  • Cells are tested negative for infectious diseases by a Mouse Essential CLEAR Panel by Charles River Animal Diagnostic Services.
  • Cells are verified to be of mouse origin and negative for interspecies contamination from human, rat, Chinese hamster, Golden Syrian hamster, and nonhuman primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
  • Cells are negative for mycoplasma contamination.

Features and Benefits

Replic renal cells have unqiue myofibroblastic features such as high TGFβ and aide in the understanding of kidney fibrosis, renal anemia, and molecular chronic kidney disease pathology.

Preparation Note

Replic cells should be stored in liquid nitrogen. The cells can be cultured for at least 10 passages without significantly affecting cell marker expression and function.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@milliporesigma.com.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCC177
description

-

description

TS/A mouse mammary adenocarcinoma cell line is a well characterized and highly published model for breast cancer research.

biological source

mouse

biological source

mouse

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

shipped in

liquid nitrogen

shipped in

ambient

relevant disease(s)

cancer

relevant disease(s)

-

storage temp.

−196°C

storage temp.

-140 to -196°C

Quality Level

100

Quality Level

-


Storage Class

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library