Skip to Content
Merck

SCC587

3dGRO® Human Liver Organoids (MNFBQ01LI (63-65))

Normal, Liver, Age: 60, Male, Mutation: None

Synonym(s):

Hepatic PDOs, Hepatocyte Organoids, Human Hepatic Organoids, Human Liver PDOs

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
NACRES:
NA.51
eCl@ss:
32040503
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

human liver

packaging

vial of ≥1500 organoids/vial vial

manufacturer/tradename

Millipore

gender

male

technique(s)

cell culture: suitable

application(s)

3D cell culture

control

liver

relevant disease(s)

(normal)

storage temp.

-140 to -196°C

General description

Patient derived organoids (PDO) are tissue derived, novel in vitro 3D cell models that recapitulate some key features of the respective organ, thus representing a more in vivo-like alternative to traditional 2D cell cultures. In addition, PDOs offer possible means to reduce animal model use. PDO maybe derived from resections or biopsy of adult tissue and are phenotypically and genetically stable in culture. PDO are an ideal platform for drug screening, disease modeling and tumor immunity studies, and may also be modified by gene editing.

The 3dGRO® human liver organoid biobank is a collection of tissue-derived normal non-diseased human liver organoids from numerous patients. The biobank contains liver PDOs that are highly viable and express high levels of albumin, HNF4-a, FOXA2, SOX17, CK7, CK19, E-Cadherin, PCK2 along with important cytochrome P450 (CYP) enzymes activities (Cyp3A4, Cyp2C9, Cyp2B6, Cyp2D6, Cyp1A2).

Cell Line Characteristics

  • Sex: Male
  • Age: 60 Years
  • Race: Asian (Indian)
  • Organ: Liver
  • Disease: Normal
  • Patient History: None

Application

  • Viability: >1500 viable organoids/vial
  • Organoid Growth: Pass
  • Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
  • Mycoplasma Contamination: Negative
  • STR Profile: Pass

Features and Benefits

Cryopreserved patient derived esophageal cancer organoids (PDOs)

Preparation Note

Store in liquid nitrogen. The organoids can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Legal Information

3dGRO® is a trademark of Merck KGaA, Darmstadt, Germany.
3dGRO® organoids were derived utilizing HUB Organoid Technology.

Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/pdouse. This product may not be re-engineered or copied; used to make derivatives or modifications; used to develop, test, or manufacture a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact organoids@milliporesigma.com.
3dGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

This product is regulated in France when intended for scientific purposes, including import, export, and conservation activities (Articles L.1211-1, paragraph 2, and L.1243-4 of the French Public Health Code). The purchaser (i.e. end user) is required to obtain an import authorization from the France Ministry responsible for Research (as referenced in Article L.1245-5-1 II of the French Public Health Code) or a conservation authorization from the same ministry (as referenced in Article L.1243-4 of the French Public Health Code). By placing an order for this product, you confirm that you have obtained the appropriate import or conservation authorization for the product. For more information regarding the use of human biological samples for research purposes in France, please contact the relevant French authorities.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library