Skip to Content

ST1069

PhosphoDetect Anti-Paxillin (pSer¹⁷⁸) Rabbit pAb

liquid, Calbiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

mouse, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

storage temp.

2-8°C

General description

Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~68 kDa paxillin protein phosphorylated at Ser178.
Paxillin is a focal adhesion adaptor protein associated with th establishment of focal adhesions, cell signaling via integrins, and cell motility. The N-terminus of paxillin contains a proline-rich region for SH3 and SH2 domain binding. The N-terminus also contains five copies of the LD motif for association of actin binding proteins. The C-terminus contains LIM domains and the focal adhesion targeting sequence. Paxillin is believed to function as an adaptor protein that facilitates assembly of multi-protein complexes and coordinates signaling via integrins. Tyrosine and serine phosphorylation of paxillin is associated with the establishment of cytoskeletal membrane attachments. Phosphorylation by JNK at Ser178 is necessary for normal cell motility.
Recognizes the ~68 kDa paxillin protein phosphorylated at Ser178.
This PhosphoDetect Anti-Paxillin (pSer¹⁷⁸) Rabbit pAb is validated for use in Immunoblotting for the detection of Paxillin (pSer¹⁷⁸).

Immunogen

Human
a synthetic phosphopeptide corresponding to amino acids surrounding the Ser¹⁷⁸ phosphorylation site of human paxillin

Application

Immunoblotting (1:1000-1:5000)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In Tris-citrate/phosphate, pH 7.0-8.0.

Preparation Note

Following initial use aliquot and freeze (-20°C) for long-term storage. Avoid freeze/thaw cylces.

Analysis Note

Positive Control
NIH3T3 cells plated on fibronectin

Other Notes

Bellis, S.L., et al. 1997. Biochem. J.325, 375.
Schaller, M.D. 2001. Oncogene20, 6459.
Huang, C., et al. 2003. Nature424, 219.
Does not recognize unphosphorylated paxillin. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Highly Toxic (H)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library