Skip to Content
Merck

PG9H9BB3873D

PrecisePEG

Azido-PEG8-Hydrazide

Synonym(s):

1-azido-3,6,9,12,15,18,21,24-octaoxaheptacosane-27-hydrazonic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C19H39O9N5
CAS Number:
Molecular Weight:
481.55
MDL number:
Product PG9H9BB3873D is not currently sold in your country.


Inchi Code

1S/C19H39N5O9/c20-23-19(25)1-3-26-5-7-28-9-11-30-13-15-32-17-18-33-16-14-31-12-10-29-8-6-27-4-2-22-24-21/h1-18,20H2,(H,23,25)

Inchi Key

XSHZSHNFUKYUOV-UHFFFAOYSA-N

Shipping Temperature

ambient

Purity

96%

Storage temp.

-20

Country of Origin

CN

Physical Form

Solid



hstatements

pcodes



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library