Skip to Content
Merck

03378

Chloride atomic spectroscopy standard concentrate 10.00 g Cl-

10.00 g/L, for 1 l standard solution, analytical standard

Synonym(s):

Hydrochloric acid solution, Hydrogen chloride solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HCl
CAS Number:
Molecular Weight:
36.46
UNSPSC Code:
41116107
PubChem Substance ID:
MDL number:
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

vapor pressure

190 mmHg ( 25 °C)

shelf life

limited shelf life, expiry date on the label

packaging

ampule of

concentration

10.00 g/L

bp

100 °C (lit.)

solubility

H2O: soluble

format

single component solution

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Preparation Note

prepared with HCl and H2O


Pictograms

Corrosion

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library