Skip to Content
Merck

08552

3-Methylcrotonaldehyde

analytical standard

Synonym(s):

3,3-Dimethylacrolein, 3,3-Dimethylacrylaldehyde, NSC 149164, Prenal, Senecialdehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3)2C=CHCHO
CAS Number:
Molecular Weight:
84.12
UNSPSC Code:
85151701
NACRES:
NA.24
PubChem Substance ID:
EC Number:
203-527-6
Beilstein/REAXYS Number:
1734740
MDL number:
eCl@ss:
32150211
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

vapor pressure

7 mmHg ( 20 °C)

assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.462

bp

133-135 °C (lit.)

density

0.878 g/mL at 20 °C, 0.872 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

[H]C(=O)\C=C(\C)C

InChI

1S/C5H8O/c1-5(2)3-4-6/h3-4H,1-2H3

InChI key

SEPQTYODOKLVSB-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under 3-Methylcrotonaldehyde


Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class

3 - Flammable liquids

Flash Point (°F)

91.4 °F - closed cup

Flash Point (°C)

33 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

107-86-8

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library