Skip to Content
Merck

Skip To

09809

Boc-allyl-Gly-OH (dicyclohexylammonium) salt

≥98.0% (TLC)

Synonym(s):

(S)-2-(Boc-amino)-4-pentenoic acid (dicyclohexylammonium) salt, Boc-L-2-allylglycine (dicyclohexylammonium) salt

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C10H17NO4 · C12H23N
CAS Number:
Molecular Weight:
396.56
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
eCl@ss:
32150313


Quality Segment

assay

≥98.0% (TLC)

form

powder

color

colorless to white

mp

117-123 °C

application(s)

peptide synthesis

SMILES string

C1CCC(CC1)NC2CCCCC2.CC(C)(C)OC(=O)N[C@@H](CC=C)C(O)=O

InChI

1S/C12H23N.C10H17NO4/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-5-6-7(8(12)13)11-9(14)15-10(2,3)4/h11-13H,1-10H2;5,7H,1,6H2,2-4H3,(H,11,14)(H,12,13)/t;7-/m.0/s1

InChI key

VMCGMPITVQIMGK-ZLTKDMPESA-N



Storage Class

13 - Non Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters