Skip to Content
Merck

Skip To

22018

L-Carnitine inner salt

≥99.0% (NT)

Synonym(s):

(−)-(R)-3-Hydroxy-4-(trimethylammonio)butyrate, Vitamin BT

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)3N+CH2CH(OH)CH2COO-
CAS Number:
Molecular Weight:
161.20
EC Number:
208-768-0
UNSPSC Code:
12352200
MDL number:
Beilstein/REAXYS Number:
4292315
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99.0% (NT)

optical activity

[α]20/D −31±1°, c = 10% in H2O

ign. residue

≤0.1% (as SO4)

loss

≤0.1% loss on drying

mp

197-212 °C (lit.)

solubility

H2O: 0.1 g/mL at 20 °C, clear, colorless

cation traces

Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Na: ≤100 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Zn: ≤5 mg/kg

storage temp.

2-8°C

SMILES string

C[N+](C)(C)C[C@H](O)CC([O-])=O

InChI

1S/C7H15NO3/c1-8(2,3)5-6(9)4-7(10)11/h6,9H,4-5H2,1-3H3/t6-/m1/s1

InChI key

PHIQHXFUZVPYII-ZCFIWIBFSA-N

Biochem/physiol Actions

Carnitine is a quaternary amine that occurs naturally in most mammalian tissue. It is present in relatively high concentrations in skeletal muscle and heart where it is involved in regulating energy metabolism. It shifts glucose metabolism from glycolysis to glycogen storage and enhances the transport of long chain fatty acids into the mitochondria where they are oxidized for energy production.

Other Notes

Review. Carnitine and its role in fatty acid metabolism




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library