Skip to Content
Merck

57720

4-Iodoanisole

purum, ≥96.0% (GC)

Synonym(s):

4-Iodophenyl methyl ether, 4-Methoxyiodobenzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
IC6H4OCH3
CAS Number:
Molecular Weight:
234.03
EC Number:
211-798-7
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
1906692
MDL number:
eCl@ss:
32160703
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥96.0% (GC)

bp

237 °C/726 mmHg (lit.)

mp

50-52 °C, 50-53 °C (lit.)

SMILES string

COc1ccc(I)cc1

InChI

1S/C7H7IO/c1-9-7-4-2-6(8)3-5-7/h2-5H,1H3

InChI key

SYSZENVIJHPFNL-UHFFFAOYSA-N

Other Notes

This product has been replaced by I7608-ALDRICH | 4-Iodoanisole 98%




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library