Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Linear Formula:
IC6H4CH2OH
CAS Number:
Molecular Weight:
234.03
EC Number:
225-933-2
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
2079487
MDL number:
Pricing and availability is not currently available.
grade
purum
assay
≥99.0% (GC)
mp
89-92 °C
SMILES string
OCc1ccccc1I
InChI
1S/C7H7IO/c8-7-4-2-1-3-6(7)5-9/h1-4,9H,5H2
InChI key
WZCXOBMFBKSSFA-UHFFFAOYSA-N
Storage Class
13 - Non Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.