Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Please contact Customer Service for Availability | 16 200,00 CZK |
About This Item
product line
Assurance GDS®
manufacturer/tradename
BioControl Systems
technique(s)
PCR: suitable
application(s)
food and beverages
pathogen testing
compatibility
for use with GDS Rotor-Gene®
storage temp.
room temp
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description BioControl Systems, for use with GDS Rotor-Gene® | description BioControl, For use with GDS | description BioControl Systems, for use with GDS Rotor-Gene® | description BioControl Systems, for use with GDS Rotor-Gene® |
| manufacturer/tradename BioControl Systems | manufacturer/tradename BioControl Systems 73083 | manufacturer/tradename BioControl Systems | manufacturer/tradename BioControl Systems |
| technique(s) PCR: suitable | technique(s) PCR: suitable | technique(s) PCR: suitable | technique(s) PCR: suitable |
| storage temp. room temp | storage temp. room temp | storage temp. room temp | storage temp. room temp |
| application(s) food and beverages | application(s) food and beverages | application(s) food and beverages | application(s) food and beverages |
| compatibility for use with GDS Rotor-Gene® | compatibility for use with GDS Rotor-Gene | compatibility for use with GDS Rotor-Gene® | compatibility for use with GDS Rotor-Gene® |
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



