Skip to Content
Merck

82811

Pyridine N-oxide

purum, ≥97.0% (NT)

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H5NO
CAS Number:
Molecular Weight:
95.10
EC Number:
211-774-6
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
105257
MDL number:
eCl@ss:
32160702
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥97.0% (NT)

impurities

≤2% water

bp

270 °C (lit.)

mp

62-67 °C

storage temp.

2-8°C

SMILES string

[O-][n+]1ccccc1

InChI

1S/C5H5NO/c7-6-4-2-1-3-5-6/h1-5H

InChI key

ILVXOBCQQYKLDS-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library