About This Item
grade
analytical standard
Quality Segment
assay
≥99.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
mp
224-226 °C (lit.)
application(s)
cleaning products
cosmetics
food and beverages
personal care
format
neat
SMILES string
Oc1ccc(cc1)C3(c2ccc(O)cc2)c4ccccc4-c5ccccc35
InChI
1S/C25H18O2/c26-19-13-9-17(10-14-19)25(18-11-15-20(27)16-12-18)23-7-3-1-5-21(23)22-6-2-4-8-24(22)25/h1-16,26-27H
InChI key
YWFPGFJLYRKYJZ-UHFFFAOYSA-N
General description
Application
- Human breast milk using gas chromatography-tandem mass spectrometry (GC-MS/MS).
- Wastewaters and surface water using gas chromatography-mass spectrometry (GC-MS).
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
WGK
WGK 2
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
3236-71-3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

