Skip to Content
Merck

HALF-1

Isotopic Certified Reference Material of natural hafnium

certified reference material

Select a Size

Change View

About This Item

NACRES:
NA.24
eCl@ss:
32129103
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

manufacturer/tradename

National Research Council of Canada (NRC)

application(s)

food and beverages

format

single component solution

storage temp.

2-8°C

General description

Certified values for isotope amount ratios, isotopic abundances, and atomic weight of hafnium have been established for this isotopic reference material

Features and Benefits

NRC Canada’s certified reference materials (CRMs) are traceable to the International System of Units, and conform to the requirements of ISO/IEC 17025 and ISO 17034.For specific values, as well as additional information regarding stability and storage information, please view the Certificate of Analysis.With NRC Canada’s CRMs, you gain access to products backed by extensive research and expertise, fostering trust and consistency in your results. Elevate your laboratory’s accuracy and credibility with NRC Canada’s certified reference materials.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

For more information, please refer to the Certificate of Analysis or Safety Data Sheet
Sales restrictions may apply.


Pictograms

Corrosion

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library