Skip to Content
Merck

Skip To

PHI90001

D-Ser(8)-Liraglutide

GLP-1 Agonist Impurity, analytical standard, Numbering in the name as per API

Synonym(s):

D-Ser(14)-Liraglutide [GLP-1 numbering], H-His-Ala-Glu-Gly-Thr-Phe-Thr-D-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-Gln-Ala-Ala-Lys(γ-Glu-palmitoyl)-Glu-Phe-Ile-Ala-Trp-Leu-Val-Arg-Gly-Arg-Gly-OH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
Please contact Customer Service for Availability
4 600,00 CZK

About This Item

Empirical Formula (Hill Notation):
C172H265N43O51
Molecular Weight:
3751.20
UNSPSC Code:
41116107
eCl@ss:
25140904

4 600,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

API family

liraglutide, liraglutide, liraglutide, liraglutide

form

powder, solid

technique(s)

HPLC: suitable, LC/MS: suitable, liquid chromatography (LC): suitable, mass spectrometry (MS): suitable

color

white to off-white

format

neat

storage temp.

−20°C

SMILES string

OC[C@H](NC([C@@H](NC([C@@H](NC([C@@H](NC([C@H](NC([C@@H](NC([C@@H](NC([C@@H](NC(CNC([C@@H](NC([C@@H](NC([C@@H](N)CC1=CNC=N1)=O)C)=O)CCC(O)=O)=O)=O)[C@@H](C)O)=O)CC2=CC=CC=C2)=O)[C@@H](C)O)=O)CO)=O)CC(O)=O)=O)C(C)C)=O)CO)=O)C(N[C@H](C(N[C@H](C(N[C@H](C(NCC(N[C@H](C(N[C@H](C(N[C@H](C(N[C@H](C(N[C@@H](CCC(O)=O)C(N[C@@H](CC3=CC=CC=C3)C(N[C@H](C(N[C@@H](C)C(N[C@H](C(N[C@H](C(N[C@H](C(N[C@@H](CCCNC(N)=N)C(NCC(N[C@@H](CCCNC(N)=N)C(NCC(O)=O)=O)=O)=O)=O)C(C)C)=O)CC(C)C)=O)CC4=CNC5=CC=CC=C45)=O)=O)[C@H](CC)C)=O)=O)=O)CCCCNC(CC[C@H](NC(CCCCCCCCCCCCCCC)=O)C(O)=O)=O)=O)C)=O)C)=O)CCC(N)=O)=O)=O)CCC(O)=O)=O)CC(C)C)=O)CC6=CC=C(C=C6)O)=O

General description

The analytical standard of D-Ser(8)-Liraglutide is a stereoisomeric impurity of Liraglutide in which the L-Serine residue at position 8, API numbering (and position 14 in GLP-1 numbering) is replaced by its D-enantiomer (D-Ser). This single chiral inversion, two residues from the N-terminus, can arise during solid-phase peptide synthesis (SPPS) through incomplete chiral control or racemisation under acidic/basic coupling conditions. The compound retains the full 31-residue backbone of Liraglutide and carries the characteristic palmitoyl–γGlu–Lys26 fatty acid conjugate responsible for albumin binding and extended half-life. However, the D-amino acid substitution is expected to locally distort the α-helical secondary structure, potentially altering GLP-1 receptor binding affinity and metabolic stability.

Application

Used as an analytical standard for the identification and semi-quantification of the D-Ser-8 stereoisomeric impurity in Liraglutide API by reversed-phase HPLC, chiral chromatography, and LC-MS/MS methods.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHI90004PHI90002SML3925
description

GLP-1 Agonist Impurity, analytical standard, Numbering in the name as per API

description

GLP-1 Agonist Impurity, analytical standard, Numbering in the name as per API

description

GLP-1 Agonist Impurity, analytical standard, Numbering in the name as per API

description

≥95% (HPLC)

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

-

form

powder, solid

form

powder

form

solid

form

(Powder or Lyophilized powder or film)

format

neat

format

neat

format

neat

format

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

storage temp.

-10 to -25°C

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

technique(s)

liquid chromatography (LC): suitable, mass spectrometry (MS): suitable, LC/MS: suitable, HPLC: suitable

technique(s)

liquid chromatography (LC): suitable, HPLC: suitable, mass spectrometry (MS): suitable, LC/MS: suitable

technique(s)

-

technique(s)

-


Pictograms

Health hazard

Signal Word

Warning

Hazard Codes

Hazard Classifications

Repr. 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library