Skip to Content
Merck

Skip To

PMB3003

L-Arginine monohydrochloride

98.5-101%, ISOM8™, suitable for cell culture, from non-animal source

Synonym(s):

L-Arginine monohydrochloride, (S)-(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride, (S)-(+)-Arginine hydrochloride, (S)-(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
Please contact Customer Service for Availability
11 100,00 CZK
Please contact Customer Service for Availability
33 300,00 CZK
Please contact Customer Service for Availability
49 900,00 CZK

About This Item

Empirical Formula (Hill Notation):
C6H14N4O2 · HCl
CAS Number:
Molecular Weight:
210.66
UNSPSC Code:
12352200
Beilstein/REAXYS Number:
3631658
MDL number:

11 100,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

non-animal source

Quality Segment

agency

meets EP, meets JP, meets USP, meets requirements for USP (testing)

product line

ISOM8™

assay

98.5-101%

form

crystalline powder

optical activity

[α]/D 22.1-22.9°, c = 6 M in HCl (8 in 6 M HCl)

technique(s)

cell culture | mammalian: suitable

color

white

mp

226-230 °C

functional group

(Guanidine), amine, carboxylic acid

storage temp.

room temp

SMILES string

Cl[H].N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H/t4-;/m0./s1

InChI key

KWTQSFXGGICVPE-WCCKRBBISA-N

General description

L-Arginine Monohydrochloride is the hydrochloride salt of L-Arginine, an essential, basic amino acid. As a cationic amino acid, L-Arginine contains a guanidino group in its side chain, which gives it a strong positive charge at physiological pH and contributes to its roles in various biochemical and cellular processes. The hydrochloride form enhances the compound′s solubility and stability, making it suitable for a wide range of aqueous applications.

Biochem/physiol Actions

Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.

Features and Benefits

Features:

1. Qualified Raw Materials: Products that meet the stringent requirements of the Elevated Assurance Program, ensuring compliance with regulatory standards.
2. Comprehensive Documentation: Includes essential documents such as certificates, impurity profiles, and declarations to support audits and risk assessments.
3. M-Clarity™ Quality Attributes: Products are classified within the MQ300 quality segment or higher, demonstrating superior quality assurance. https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/m-clarity-program
4. Confirmed Process Controls: Established processes that ensure consistency and reliability in manufacturing.
5. Enhanced Change Notification: Timely updates on any changes to product specifications or processes, ensuring transparency and compliance.
6. Freely Available Dossiers: Access to robust, ready-to-present product dossiers that include:
  • Site quality assessment
  • ISO 9001 certificate
  • Detailed product specifications
  • Relevant declarations/statements (e.g., animal origin content, BSE/TSE, nitrosamine)


Benefits:


1. Regulatory Compliance: Helps you meet compliance standards effectively, reducing the risk of regulatory issues.
2. Audit Readiness: Instills confidence during audits with comprehensive documentation and confirmed process controls, demonstrating your commitment to high-quality products.
3. Operational Transparency: Supports supply chain transparency, allowing you to track and verify the quality of raw materials.
4. Enhanced Support: Provides exceptional quality-related information and tailored assistance to meet your specific needs.
5. Risk Mitigation: Facilitates thorough risk assessments with detailed documentation, helping you manage potential compliance challenges.
6. Informed Decision-Making: Access to clear and comprehensive product information enables better decision-making and enhances operational efficiency.

Legal Information

ISOM8 is a trademark of Sigma-Aldrich Co. LLC
M-CLARITY is a trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1454BP6941042601
description

98.5-101%, ISOM8™, suitable for cell culture, from non-animal source

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

British Pharmacopoeia (BP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

form

crystalline powder

form

-

form

crystalline

form

powder

assay

98.5-101%

assay

-

assay

-

assay

-

functional group

amine, (Guanidine), carboxylic acid

functional group

-

functional group

-

functional group

-

technique(s)

cell culture | mammalian: suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

product line

ISOM8™

product line

-

product line

-

product line

-

storage temp.

room temp

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

-


Still not finding the right product?

Explore all of our products under L-Arginine monohydrochloride


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library