Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12352200
NACRES:
NA.21
Pricing and availability is not currently available.
grade
analytical standard
assay
>95%
mol wt
(11.7 kD)
manufacturer/tradename
InBio
format
neat
storage temp.
−20°C
General description
Source: E. coli Purification: Purified from E. coli culture by multi-step chromatography.
Formulation: 0.2M Arginine, 1X PBS, pH 7.4. Filtered through 0.22µm filter.
Notes: (1) rPru du 3 has an N-terminal 8xHis-tag.
(2) N-glycosylation motif at position 33 was mutated (N33Q).
Formulation: 0.2M Arginine, 1X PBS, pH 7.4. Filtered through 0.22µm filter.
Notes: (1) rPru du 3 has an N-terminal 8xHis-tag.
(2) N-glycosylation motif at position 33 was mutated (N33Q).
Disclaimer
For research and commercial use in vitro: not for human in vivo or therapeutic use.
Storage Class
12 - Non Combustible Liquids
WGK
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.