Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
500 mL | Please contact Customer Service for Availability | 4 840,00 CZK |
About This Item
grade
certified reference material
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
viscosity
8081 mPa.s(25.00 °C)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
mixture
General description
1 of 1
This Item | |||
|---|---|---|---|
| description viscosity 8081 mPa.s (25.00 °C) | description viscosity 29.04 mPa.s (25.00 °C) | description viscosity 250.0 mPa.s (25.00 °C) | description viscosity 389.1 mPa.s (25.00 °C) |
| format mixture | format mixture | format mixture | format mixture |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| application(s) cleaning products | application(s) cleaning products | application(s) cleaning products | application(s) cleaning products |
| manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd |
| viscosity 8081 mPa.s(25.00 °C) | viscosity 29.04 mPa.s(25.00 °C) | viscosity 250.0 mPa.s(25.00 °C) | viscosity 389.1 mPa.s(25.00 °C) |
| shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label |
Still not finding the right product?
Explore all of our products under Mineral Oil Rotational Viscosity Standard
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Asp. Tox. 1 - Skin Irrit. 2
Storage Class
10 - Combustible liquids
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


