Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
Please contact Customer Service for Availability | 10 500,00 CZK |
About This Item
10 500,00 CZK
grade
certified reference material
agency
traceable to (National Research Council of Canada (NRC))
manufacturer/tradename
National Research Council of Canada (NRC)
application(s)
food and beverages
format
neat
storage temp.
room temp
General description
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description certified reference material | description certified reference material | description certified reference material | description certified reference material |
| application(s) food and beverages | application(s) food and beverages | application(s) food and beverages | application(s) food and beverages |
| format neat | format neat | format neat | format matrix material |
| agency traceable to (National Research Council of Canada (NRC)) | agency traceable to (National Research Council of Canada (NRC)) | agency - | agency traceable to (National Research Council of Canada (NRC)) |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| storage temp. room temp | storage temp. room temp | storage temp. - | storage temp. - |
| manufacturer/tradename National Research Council of Canada (NRC) | manufacturer/tradename National Research Council of Canada (NRC) | manufacturer/tradename National Research Council of Canada (NRC) | manufacturer/tradename National Research Council of Canada (NRC) |
Still not finding the right product?
Explore all of our products under Isotopic certified reference material of vanillin
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class
11 - Combustible Solids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

