Skip to Content
Merck

Skip To

42945

(−)-Hydroxymatairesinol, mixture of epimers

≥90% (HPLC)

Synonym(s):

(3R,4R)-Dihydro-4-[hydroxy(4-hydroxy-3-methoxyphenyl)methyl]-3-[(4-hydroxy-3-methoxyphenyl)methyl]-2(3H)-furanone, 7-Hydroxymatairesinol-7-allo-Hydroxymatairesinol mixture

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C20H22O7
CAS Number:
Molecular Weight:
374.38
PubChem Substance ID:
UNSPSC Code:
12352200


assay

≥90% (HPLC)

SMILES string

COc1cc(C[C@H]2COC(=O)[C@H]2C(O)c3ccc(O)c(OC)c3)ccc1O

InChI

1S/C20H22O7/c1-25-16-8-11(3-5-14(16)21)7-13-10-27-20(24)18(13)19(23)12-4-6-15(22)17(9-12)26-2/h3-6,8-9,13,18-19,21-23H,7,10H2,1-2H3/t13-,18+,19?/m0/s1

InChI key

ZZSOKNNVDKKSDE-HDOPWBOFSA-N

Application

This product is an epimeric mixture of hydroxymatairesinol, a plant phytoestrogen, which may be used to study its proestrogenic, antioxidant and chemopreventative properties.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Pictograms

Environment

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters