Skip to Content
Merck

Skip To

44367

Lewis-a trisaccharide

≥98.0% (TLC)

Synonym(s):

α-L-Fuc-(1→4)-[β-D-Gal-(1→3)]-D-GlcNAc, Galactosyl-β-1,3-(fucosyl-α-1,4)-N-acetylglucosamine, Lea-Trisaccharide

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C20H35NO15
CAS Number:
Molecular Weight:
529.49
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
MDL number:


assay

≥98.0% (TLC)

form

solid

technique(s)

thin layer chromatography (TLC): suitable

storage temp.

−20°C

SMILES string

OC1[C@H](NC(C)=O)[C@@H](O[C@@H]([C@H](O)[C@H]2O)O[C@H](CO)[C@@H]2O)[C@H](O[C@H](O[C@H]3C)[C@@H](O)[C@H](O)[C@@H]3O)[C@@H](CO)O1

InChI

1S/C20H35NO15/c1-5-10(25)12(27)14(29)19(32-5)35-16-8(4-23)33-18(31)9(21-6(2)24)17(16)36-20-15(30)13(28)11(26)7(3-22)34-20/h5,7-20,22-23,25-31H,3-4H2,1-2H3,(H,21,24)/t5-,7+,8+,9+,10+,11-,12+,13-,14-,15+,16+,17+,18?,19-,20-/m0/s1

InChI key

CFDVGUXRLQWLJX-QGTNPELVSA-N

Biochem/physiol Actions

Lewis a trisaccharide is a fucosyloligosaccharide present in human milk and colostrum, which is important for the protection of infants against infection by pathogenic microorganisms. Lewis a trisaccharide is found on the surface of erythrocytes and is a tumor-related antigen. Used as analytical standard, enzyme substrate, in blood group typing, and antibody binding assays.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters