Skip to Content
Merck

68614

PEG 20000 12% – Tris pH 8; 100 mM solution

Synonym(s):

Crystallization solution 37/Kit-No 02528

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

pH

8.0±0.2 ( in neat)

density

1.02 g/mL at 20 °C

storage temp.

2-8°C



Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
68614-10ML-KC-F04061838040350