Skip to Content
Merck

Skip To

74384

1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt

≥98.0% (TLC)

Synonym(s):

(9Z)-9-Octadecenoic acid 1-[[[(2,3-dihydroxypropoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester monosodium salt, 1,2-Di(9Z-octadecenoyl)-sn-glycero-3-phospho-(1′-rac-glycerol) sodium salt, L-α-Phosphatidyl-DL-glycerol, dioleoyl, L-α-Phosphatidyl-DL-glycerol, dioleoyl sodium salt, DOPG-Na, PG(18:1(9Z)/18:1(9Z))

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
C42H78O10PNa
CAS Number:
Molecular Weight:
797.03
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.85


biological source

synthetic

assay

≥98.0% (TLC)

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

[Na].CCCCCCCC\C=C\CCCCCCCC(=O)OCC(COP(O)(=O)OCC(O)CO)OC(=O)CCCCCCC\C=C\CCCCCCCC

InChI

1S/C42H79O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-41(45)49-37-40(38-51-53(47,48)50-36-39(44)35-43)52-42(46)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h17-20,39-40,43-44H,3-16,21-38H2,1-2H3,(H,47,48);/q;+1/p-1/b19-17-,20-18-;/t39?,40-;/m1./s1

InChI key

IUVFCFQZFCOKRC-IPKKNMRRSA-M

General description

The lipid DOPG has significant influence on multidrug transport. The DOPG membrane content can alter substrate transport of small multidrug transporters (SMR) proteins by an order of magnitude.

Biochem/physiol Actions

Remodeling of diacylphosphatidylglycerols by transesterification was studied in a phosphatidylglycerol-deficient microbe grown with exogenous DOPG.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

PG(18:1(9Z)/18:1(9Z))


Still not finding the right product?


Storage Class

13 - Non Combustible Solids

WGK

WGK 3

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters