Skip to Content
Merck

Skip To

A0153

ADA disodium salt

Synonym(s):

N-(2-Acetamido)-2-iminodiacetic acid, N-(Carbamoylmethyl)iminodiacetic acid

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
H2NCOCH2N(CH2CO2Na)2
CAS Number:
Molecular Weight:
234.12
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:


assay

≥98% H2SO4 basis (dry, titration)

form

crystalline powder

useful pH range

6.0-7.2

pKa (25 °C)

6.6 (Free acid)

solubility

water: 0.5 g/mL, clear, colorless

application(s)

diagnostic assay manufacturing

SMILES string

[Na+].[Na+].NC(=O)CN(CC([O-])=O)CC([O-])=O

InChI

1S/C6H10N2O5.2Na/c7-4(9)1-8(2-5(10)11)3-6(12)13;;/h1-3H2,(H2,7,9)(H,10,11)(H,12,13);;/q;2*+1/p-2

InChI key

UFJHJSRPIBTMAS-UHFFFAOYSA-L



Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters