Skip to Content
Merck

Skip To

A1355

Anti-Archvillin antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, PBS solution

Synonym(s):

Anti-SVIL, Anti-Supervillin (p205/p250), Anti-Supervillin isoform 2, SmAV (Smooth muscle Archvillin)

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IHC (p), WB
Citations:
3


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

PBS solution

mol wt

antigen ~250 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 20-40 μg/mL using heat-retrieved tissue sections from human heart by biotin/ExtrAvidin-Peroxidase, indirect immunofluorescence: 20-40 μg/mL using differentiated rat L8 cells, western blot: 2.5-5 μg/mL using whole extract of differentiated mouse C2 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SVIL(6840)
mouse ... Svil(225115)
rat ... Svil(361256)

General description

Archvillin is a muscle-specific isoform of Supervillin. Archvillin binds F-actin and is believed to be involved in myogenesis and muscle cell migration. Archvillin binds ERK and modulates ERK signaling in smooth cells of the aortas of ferrets.
Rabbit anti-archvillin antibody binds to human, mouse, and rat archvillin.

Immunogen

synthetic peptide corresponding to amino acid residues 650-666 of human archvillin. The corresponding sequence is identical in mouse.

Application

Rabbit anti-archvillin antibody can be used for immunohistochemistry at 20-40 μg/mL using heat-retrieved tissue sections from human heart by biotin/ExtrAvidin-Peroxidase. The antibody can also be used for indirect immunofluorescence at 20-40 μg/mL using differentiated rat L8 cells. Furthermore, the antibody is suitable for use in western blot (2.5-5 μg/mL using whole extract of differentiated mouse C2 cells) assays.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters