Skip to Content
Merck

Skip To

A1479

Angiopoietin-4 human

recombinant, expressed in NSO cells, ≥85% (SDS-PAGE), lyophilized powder

Synonym(s):

ANG-4

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
51111800
MDL number:
eCl@ss:
32150313
Form:
lyophilized powder
Assay:
≥85% (SDS-PAGE)
Biological source:
human
Recombinant:
expressed in NSO cells
Mol wt:
apparent mol wt 60-100 kDa by SDS-PAGE (reducing), calculated mol wt 57 kDa


biological source

human

recombinant

expressed in NSO cells

assay

≥85% (SDS-PAGE)

form

lyophilized powder

mol wt

apparent mol wt 60-100 kDa by SDS-PAGE (reducing), calculated mol wt 57 kDa

storage condition

avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... ANGPT4(51378)

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline, pH 6.8, containing 1.25 mg bovine serum albumin.

Analysis Note

The biological activity is measured by its ability to compete with biotinylated recombinant human ANG-2 for binding to immobilized Tie-2/Fc in an ELISA.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters