Skip to Content
Merck

Skip To

A252

AGN 192403 hydrochloride

solid

Synonym(s):

(2-endo, 3-exo)-3-(Methylethyl)-bicyclo[2.2.1]heptan-2-amine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H19N · HCl
CAS Number:
Molecular Weight:
189.73
UNSPSC Code:
12352200
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

color

off-white

solubility

H2O: 20 mg/mL

SMILES string

Cl.CC(C)[C@@H]1[C@H]2CC[C@H](C2)[C@H]1N

Gene Information

human ... NISCH(11188)

Biochem/physiol Actions

I1 imidazoline receptor ligand.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library