Skip to Content
Merck

A2956

Ammonium tartrate dibasic

BioXtra

Synonym(s):

L-(+)-Tartaric acid diammonium salt, Diammonium tartrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(NH4)2C4H4O6
CAS Number:
Molecular Weight:
184.15
NACRES:
NA.25
PubChem Substance ID:
eCl@ss:
39021923
UNSPSC Code:
12161700
EC Number:
221-618-9
MDL number:
Beilstein/REAXYS Number:
6120352
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioXtra

assay

≥98% (titration)

form

powder

impurities

≤0.0005% Phosphorus (P), ≤0.1% Insoluble matter

ign. residue

≤0.1%

pH

5.0-7.0

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤0.05%, sulfate (SO42-): ≤0.1%

cation traces

Al: ≤0.0005%, Ca: ≤0.0010%, Cu: ≤0.0005%, Fe: ≤0.0005%, K: ≤0.005%, Mg: ≤0.0005%, Na: ≤0.005%, Pb: ≤0.001%, Zn: ≤0.0005%

storage temp.

room temp

SMILES string

N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6.2H3N/c5-1(3(7)8)2(6)4(9)10;;/h1-2,5-6H,(H,7,8)(H,9,10);2*1H3/t1-,2-;;/m1../s1

InChI key

NGPGDYLVALNKEG-OLXYHTOASA-N



Still not finding the right product?

Explore all of our products under Ammonium tartrate dibasic


Storage Class

13 - Non Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library