Skip to Content
Merck

Skip To

A8906

Apigenin 7-O-neohesperidoside

Synonym(s):

Rhoifolin, Rhoifoloside

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C27H30O14
CAS Number:
Molecular Weight:
578.52
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
EC Number:
241-335-4
MDL number:
eCl@ss:
39020601


form

solid

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2Oc3cc(O)c4C(=O)C=C(Oc4c3)c5ccc(O)cc5)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C27H30O14/c1-10-20(32)22(34)24(36)26(37-10)41-25-23(35)21(33)18(9-28)40-27(25)38-13-6-14(30)19-15(31)8-16(39-17(19)7-13)11-2-4-12(29)5-3-11/h2-8,10,18,20-30,32-36H,9H2,1H3/t10-,18+,20-,21+,22+,23-,24+,25+,26-,27+/m0/s1

InChI key

RPMNUQRUHXIGHK-PYXJVEIZSA-N

Application

Apigenin 7-O-neohesperidoside is a flavonoid found in plants such as Paeonia suffruticosa and Buddleja albiflora. Apigenin 7-O-neohesperidoside is useful as a reference compound in procedures for the separation and detection of flavonoids.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Still not finding the right product?

Explore all of our products under Apigenin 7-O-neohesperidoside


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters