Skip to Content
Merck

Skip To

APREST93046

PrEST Antigen DUSP2

Prestige Antigens Powered by Atlas Antibodies, buffered aqueous solution

Synonym(s):

PAC-1

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352202


recombinant

expressed in E. coli

assay

>80% (SDS-PAGE)

form

buffered aqueous solution

mol wt

predicted mol wt 23 kDa

purified by

immobilized metal affinity chromatography (IMAC)

concentration

≥0.5 mg/mL

immunogen sequence

GPTAVYFLRGGFDGFQGCCPDLCSEAPAPALPPTGDKTSRSDSRAPVYDQGGPV

Ensembl | human accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... DUSP2(1844)

General description

Recombinant protein fragment of Human DUSP2 with N-terminal His6ABP (ABP = Albumin Binding Protein derived from Streptococcal Protein G) fusion tag.    

Application

Blocking agent and positive assay control using corresponding antibodies.

Physical form

Solution in phosphate-buffered saline and 1M Urea, pH 7.4

Preparation Note

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user.

Other Notes

Corresponding Antibody HPA071920.

Legal Information

Prestige Antigens is a trademark of Sigma-Aldrich Co. LLC


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters