Skip to Content
Merck

B0436

Anti-BAF57 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Mammalian Chromatin Remodeling Complex BRG1-associated Factor 57, Anti-SWI/SNF Related, Matrix Associated, Actin Dependent Regulator of Chromatin, Subfamily E, Member 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
MDL number:
eCl@ss:
32160702
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 57 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

indirect immunofluorescence: 5-10 μg/mL using mouse NIH-3T3 cells fixed with paraformaldehyde-triton, microarray: suitable, western blot: 0.25-0.5 μg/mL using HeLa nuclear cell extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SMARCE1(6605)
mouse ... Smarce1(57376)

Immunogen

synthetic peptide corresponding to amino acids 394-411 of human BAF57. The sequence is conserved in mouse.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.


Still not finding the right product?

Try our Product Selector Tool to narrow your options




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library