Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
form
solid
optical activity
[α]25/D −102.7°, c = 0.9 in methanol(lit.)
color
white
solubility
aqueous base: unstable, water and ethanol: soluble (Solutions should be freshly prepared.)
storage temp.
2-8°C
SMILES string
[Br-].C[N+]1(C)CCc2cc3OCOc3cc2[C@H]1[C@@H]4OC(=O)c5c6OCOc6ccc45
InChI
1S/C21H20NO6.BrH/c1-22(2)6-5-11-7-15-16(26-9-25-15)8-13(11)18(22)19-12-3-4-14-20(27-10-24-14)17(12)21(23)28-19;/h3-4,7-8,18-19H,5-6,9-10H2,1-2H3;1H/q+1;/p-1/t18-,19+;/m0./s1
InChI key
BWXCECYGGMGBHD-GRTNUQQKSA-M
Gene Information
Biochem/physiol Actions
Disclaimer
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

