Skip to Content
Merck

Skip To

B1383

Benzalkonium chloride

semisolid

Synonym(s):

Alkylbenzyldimethylammonium chloride, Alkyldimethylbenzylammonium chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C6H5CH2N(CH3)2RCl (R=C8H17 to C18H37)
CAS Number:
EC Number:
264-151-6
UNSPSC Code:
12352200
MDL number:
Beilstein/REAXYS Number:
4062599
eCl@ss:
32150319
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

cationic

form

semisolid

SMILES string

[Cl-].CCCCCCCCCCCCCC[N+](C)(C)Cc1ccccc1

InChI

1S/C19H34N.ClH/c1-4-5-6-7-8-9-10-14-17-20(2,3)18-19-15-12-11-13-16-19;/h11-13,15-16H,4-10,14,17-18H2,1-3H3;1H/q+1;/p-1

InChI key

TTZLKXKJIMOHHG-UHFFFAOYSA-M



Still not finding the right product?

Explore all of our products under Benzalkonium chloride


Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library