Skip to Content
Merck

Skip To

B5154

Betaine monohydrate

Synonym(s):

Betaine, (Carboxymethyl)trimethylammonium inner salt, Oxyneurine, Trimethylglycine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)3N+CH2COO-
CAS Number:
Molecular Weight:
117.15
UNSPSC Code:
12352202
Beilstein/REAXYS Number:
3537113
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

(Powder to crystal)

Quality Segment

solubility

soluble (50mg/ml)

storage temp.

2-8°C

SMILES string

C[N+](C)(C)CC([O-])=O

InChI

1S/C5H11NO2/c1-6(2,3)4-5(7)8/h4H2,1-3H3

InChI key

KWIUHFFTVRNATP-UHFFFAOYSA-N

General description

Betaine is a zwitterionic quaternary ammonium compound and is mainly present in animals, microorganisms and plants. It is one of the main constituent of several food items like wheat, shellfish, spinach and sugar beets. It is also termed as trimethylglycine, lycine, glycine betaine and oxyneurine. Betaine is a methyl derivative of glycine.

Application

Betaine is a methyl group donor that functions in the normal metabolic cycle of methionine. It is a naturally occurring choline derivative commonly ingested through diet, with a role in regulating cellular hydration and maintaining cell function.Betaine in culture medium can effectively reduce the level of ROS.

Biochem/physiol Actions

End-product of oxidative metabolism of choline, betaine is a general methyl donor, in particular in a minor pathway of methionine biosynthesis. It is used to treat homocystinuria, which is a defect in the major pathway of methionine biosynthesis.

Features and Benefits

Betaine is a modified amino acid consisting of glycine with three methyl groups serving as methyl donor for various metabolic pathways.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
W422312619621065695
description

-

description

≥98%

description

BioUltra, ≥99.0% (NT)

description

United States Pharmacopeia (USP) Reference Standard

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

-

form

(Powder to crystal)

form

-

form

-

form

-

storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

-

solubility

soluble (50mg/ml)

solubility

-

solubility

methanol: 1 M at 20 °C, clear, colorless

solubility

-


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library