Skip to Content
Merck

Skip To

BPS50030

HDAC Developer (2X)

Synonym(s):

buffers

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41105331
eCl@ss:
32160703
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

BPS Bioscience

storage temp.

-65 to -96°C

General description

HDAC assay developer is an essential reagent for signal generation in HDAC assays performed with any of BPS Bioscience′s HDAC fluorimetric substrates or assay kits. An appropriate fluorimetric substrate, comprising an acetylated lysine side chain, is first incubated with a sample containing HDAC activity (nuclear or cellular extract, purified enzyme, bead-bound immunocomplex, etc.). Deacetylation of the substrate sensitizes it so that, in a second step, treatment with the HDAC assay developer produces a fluorophore. Trichostatin A is included as an ′inhibitor stop′ for class I and II HDACs.Datasheet

Application

Study of HDAC kinetics, regulation and inhibitor sensitivity. Ideal for drug discovery and HTS applications and cell-based HDAC assays.


Datasheet and Protocols may be found here.

Preparation Note

A 2x concentrate. Product is to be used (undiluted) at 1:1 ratio with HDAC reaction.

Analysis Note

Avoid freeze/thaw cycles. Use product undiluted.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BPS53029BPS78856566328
description

-

description

-

description

-

description

-

storage temp.

-65 to -96°C

storage temp.

-65 to -96°C

storage temp.

-65 to -96°C

storage temp.

-

manufacturer/tradename

BPS Bioscience

manufacturer/tradename

BPS Bioscience

manufacturer/tradename

BPS Bioscience

manufacturer/tradename

-


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

9002-07-7

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library