Skip to Content
Merck

Skip To

C0639

(±)-3-Chloro-1,2-propanediol

Synonym(s):

α-Chlorohydrin, α-Glycerol chlorohydrin, α-Monochlorohydrin, 3-MCPD

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
ClCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
110.54
EC Number:
202-492-4
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
635684
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.04 mmHg ( 25 °C)

refractive index

n20/D 1.480 (lit.)

bp

213 °C (lit.)

density

1.322 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OCC(O)CCl

InChI

1S/C3H7ClO2/c4-1-3(6)2-5/h3,5-6H,1-2H2

InChI key

SSZWWUDQMAHNAQ-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under (±)-3-Chloro-1,2-propanediol




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library