Skip to Content
Merck

C2012

2-(p-Chlorophenyl)ethylamine

Synonym(s):

p-Chlorophenethylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

EC Number:
205-853-4
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

drug control

Új pszichoaktív anyag / New psychoactive substance (Hungary), 78/2022. (XII. 28.) BM rendelet

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

density

1.12 g/mL at 25 °C (lit.)

format

neat

storage temp.

−20°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

222.8 °F - closed cup

Flash Point (°C)

106 °C - closed cup

PPE (personal protective equipment)

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library