Skip to Content
Merck

C4483

CD152/Fc Chimera, Cytolytic from mouse

recombinant, expressed in NS.1 cells, buffered aqueous solution

Synonym(s):

CTLA4/Fc Chimera, cytolytic

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
51111800
NACRES:
NA.77
MDL number:
eCl@ss:
32160703
Form:
buffered aqueous solution
Assay:
≥99% (SDS-PAGE)
Biological source:
mouse
Recombinant:
expressed in NS.1 cells
Mol wt:
97 kDa
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

recombinant

expressed in NS.1 cells

assay

≥99% (SDS-PAGE)

form

buffered aqueous solution

mol wt

97 kDa

packaging

pkg of 1 mg

storage condition

avoid repeated freeze/thaw cycles

impurities

≤1 EU/mg protein Endotoxin level (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

The complement (C1q) and FcγR I binding sites of the Fcγ2a fragment allow the Fc portion of the fusion protein to effectively facilitate direct antibody directed cytotoxicity (ADCC) and complement directed cytotoxicity (CDC).

Biochem/physiol Actions

CD152 (CTLA4), a cell surface glycoprotein expressed at low levels on activated T cells, is a high affinity receptor for the costimulatory molecules CD80 (B7-1) and CD86 (B7-2). A related cell surface glycoprotein, CD28, binds to CD80 and CD86 with lower affinity. The soluble CD152/Fc chimeric fusion protein blocks the B7/CD28 signaling pathway by binding to CD80 and CD86.

Physical form

Solution, 0.2 μm filtered, in phosphate buffered saline, pH 7.4, with no preservative added.

Preparation Note

Purified from serum-free tissue culture supernatant

Other Notes

The extracellular domain (160 amino acids) of mouse CD152 is fused to mouse IgG2a Fc domain.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library